2-[(2,4-Dichlorobenzoyl)amino]acetic acid ≥95%
Supplier: Matrix Scientific
044756-500MG
044756-5G
044756-1G
101867-454EA
532.31
USD
101867-454
101867-450
101867-452
2-[(2,4-Dichlorobenzoyl)amino]acetic acid ≥95%
2-[(2,4-Dichlorobenzoyl)amino]acetic acid
Hazard: Irritant
Formula:
C₉H₇Cl₂NO₃ MW: 248.07 g/mol Melting Pt: 156…160 °C |
MDL Number:
MFCD00619713 CAS Number: 7554-79-2 |
Specification Test Results
Melting point | 156-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...