3,3-Bis(hydroxymethyl)-2,3-dihydro-4H-thiochromen-4-one ≥95%

Supplier: Matrix Scientific

044755-1G 044755-500MG
101867-446EA 1109.23 USD
101867-446 101867-448
3,3-Bis(hydroxymethyl)-2,3-dihydro-4H-thiochromen-4-one ≥95%
3,3-Bis(hydroxymethyl)-2,3-dihydro-4H-thiochromen-4-one

Hazard: Irritant

Formula: C₁₁H₁₂O₃S
Melting Pt: 142…145 °C
MDL Number: MFCD00139867
CAS Number: 29107-30-0

Order Now

Specification Test Results

Melting point 142-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR