6-Chloroimidazo[2,1-b][1,3]thiazole ≥95%

Supplier: Matrix Scientific

044735-1G 044735-500MG 044735-5G
101867-342EA 698.46 USD
101867-342 101867-340 101867-344
6-Chloroimidazo[2,1-b][1,3]thiazole ≥95%
6-Chloroimidazo[2,1-b][1,3]thiazole

Hazard: Irritant

Formula: C₅H₃ClN₂S
MW: 158.61 g/mol
Melting Pt: 83…84 °C
MDL Number: MFCD00173802
CAS Number: 23576-81-0

Order Now

Specification Test Results

Melting point 83-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR