Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Supplier: Matrix Scientific
044731-5G
044731-500MG
044731-1G
101867-320EA
1073.85
USD
101867-320
101867-316
101867-318
Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₉F₃N₂O₄ MW: 314.22 g/mol Melting Pt: 119…121 °C |
MDL Number:
MFCD00141006 CAS Number: 121582-55-6 |
Specification Test Results
Melting point | 119-121°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...