5-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

044723-5G
101867-280EA 64.62 USD
101867-280
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₅H₆N₄
Melting Pt: 221…223 °C
MDL Number: MFCD00114569
CAS Number: 5334-41-8

Order Now

Specification Test Results

Melting point 221-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR