2-(4-(Trifluoromethyl)phenyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

044695-5G
101867-152EA 492.31 USD
101867-152
2-(4-(Trifluoromethyl)phenyl)-4-thiazolecarboxylic acid ≥95%
2-(4-(Trifluoromethyl)phenyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃NO₂S
Melting Pt: 188…191 °C
MDL Number: MFCD05865138
CAS Number: 144061-16-5

Order Now

Specification Test Results

Melting point 188-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR