2-(4-Fluorophenyl)-3-nitropyridine ≥95%

Supplier: Matrix Scientific

044663-1G 044663-500MG 044663-5G
101866-990EA 1073.85 USD
101866-990 101866-986 101866-988
2-(4-Fluorophenyl)-3-nitropyridine ≥95%
2-(4-Fluorophenyl)-3-nitropyridine

Hazard: Irritant

Formula: C₁₁H₇FN₂O₂
Melting Pt: 94…96 °C
MDL Number: MFCD06200968
CAS Number: 168267-36-5

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR