4-(3-Nitro-2-pyridinyl)benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
044661-1G
044661-500MG
044661-5G
101866-974EA
612.31
USD
101866-974
101866-976
101866-978
4-(3-Nitro-2-pyridinyl)benzenecarbaldehyde ≥95%
4-(3-Nitro-2-pyridinyl)benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₈N₂O₃ MW: 228.21 g/mol Melting Pt: 128…130 °C |
MDL Number:
MFCD06200967 CAS Number: 847446-87-1 |
Specification Test Results
Melting point | 128-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...