2-(4-Fluorophenyl)-5-nitropyridine ≥95%

Supplier: Matrix Scientific

044659-5G 044659-1G 044659-500MG
101866-962EA 280.6 USD
101866-962 101866-964 101866-966
2-(4-Fluorophenyl)-5-nitropyridine ≥95%
2-(4-Fluorophenyl)-5-nitropyridine

Hazard: Irritant

Formula: C₁₁H₇FN₂O₂
Melting Pt: 132…136 °C
MDL Number: MFCD06200966
CAS Number: 886361-78-0

Order Now

Specification Test Results

Melting point 132-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR