4-(5-Nitro-2-pyridinyl)benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
044658-5G
044658-1G
044658-500MG
101866-960EA
1600
USD
101866-960
101866-958
101866-956
4-(5-Nitro-2-pyridinyl)benzenecarbaldehyde ≥95%
4-(5-Nitro-2-pyridinyl)benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₈N₂O₃ MW: 228.21 g/mol Melting Pt: 180…182 °C |
MDL Number:
MFCD05864831 CAS Number: 433920-97-9 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...