Pentafluorophenyl 5-bromo-2-thiophenesulfonate ≥95%

Supplier: Matrix Scientific

044653-1G 044653-500MG 044653-5G
101866-930EA 1073.85 USD
101866-930 101866-928 101866-926
Pentafluorophenyl 5-bromo-2-thiophenesulfonate ≥95%
Pentafluorophenyl 5-bromo-2-thiophenesulphonate

Hazard: Irritant

Formula: C₁₀H₂BrF₅O₃S₂
Melting Pt: 38…40 °C
MDL Number: MFCD06659553
CAS Number: 885949-67-7

Order Now

Specification Test Results

Melting point 38-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR