Methyl-6-(1H-pyrrol-1-yl)nicotinate ≥95%

Supplier: Matrix Scientific

044640-5G 044640-500MG 044640-1G
101866-852EA 556.92 USD
101866-852 101866-854 101866-856
Methyl-6-(1H-pyrrol-1-yl)nicotinate ≥95%
Methyl-6-(1H-pyrrol-1-yl)nicotinate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 125…129 °C
MDL Number: MFCD04973730
CAS Number: 852180-80-4

Order Now

Specification Test Results

Melting point 125-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR