3-(1H-Pyrrol-1-yl)-3-(3-thienyl)propanoic acid ≥95%
Supplier: Matrix Scientific
044624-5G
044624-500MG
044624-1G
101866-784EA
927.69
USD
101866-784
101866-786
101866-782
3-(1H-Pyrrol-1-yl)-3-(3-thienyl)propanoic acid ≥95%
3-(1H-Pyrrol-1-yl)-3-(3-thienyl)propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S Melting Pt: 99 °C |
MDL Number:
MFCD03787346 CAS Number: 866019-33-2 |
Specification Test Results
Melting point | 99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...