Ethyl-5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate

044617-500MG 044617-5G
101866-742EA 181.54 USD
101866-742 101866-746
Ethyl-5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl 5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃O₂
MW: 265.7000 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD00973940
CAS Number: 15001-08-8

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR