N-Hydroxy-2-phenyl-1-hydrazinecarboxamide ≥95%
Supplier: Matrix Scientific
044613-5G
044613-1G
044613-500MG
101866-720EA
427.69
USD
101866-720
101866-718
101866-722
N-Hydroxy-2-phenyl-1-hydrazinecarboxamide ≥95%
N-Hydroxy-2-phenyl-1-hydrazinecarboxamide
Hazard: Irritant
Formula:
C₇H₉N₃O₂ Melting Pt: 156…158 °C |
MDL Number:
MFCD03791220 CAS Number: 121933-76-4 |
Specification Test Results
Melting point | 156-158°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...