Methyl-4-(3,4-diaminophenoxy)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
044591-1G
044591-500MG
101866-616EA
747.69
USD
101866-616
101866-618
Methyl-4-(3,4-diaminophenoxy)benzenecarboxylate ≥95%
Methyl-4-(3,4-diaminophenoxy)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₄N₂O₃ Melting Pt: 92…95 °C |
MDL Number:
MFCD03425779 CAS Number: 24002-80-0 |
Specification Test Results
Melting point | 92-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...