3-(1,3-Benzodioxol-5-yloxy)-2-butanone ≥95%
Supplier: Matrix Scientific
044590-500MG
044590-5G
044590-1G
101866-610EA
181.54
USD
101866-610
101866-612
101866-614
3-(1,3-Benzodioxol-5-yloxy)-2-butanone ≥95%
3-(1,3-Benzodioxol-5-yloxy)-2-butanone
Hazard: Irritant
Formula:
C₁₁H₁₂O₄ Melting Pt: 30…32 °C |
MDL Number:
MFCD03425778 CAS Number: 23973-82-2 |
Specification Test Results
Melting point | 30-32°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...