Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-(5-Oxazolyl)benzoate
, Methyl 4-(1,3-oxazol-5-yl)benzoate
044585-5G
044585-500MG
044585-1G
101866-582EA
350.77
USD
101866-582
101866-584
101866-580
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate ≥95%
Methyl-4-(1,3-oxazol-5-yl)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₉NO₃ MW: 203.2 g/mol Melting Pt: 112…114 °C |
MDL Number:
MFCD03012557 CAS Number: 179057-14-8 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...