6-Hydroxy-1,4-dimethyl-5-(morpholinomethyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
044574-500MG
044574-1G
044574-5G
101866-528EA
806.15
USD
101866-528
101866-524
101866-526
6-Hydroxy-1,4-dimethyl-5-(morpholinomethyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
6-Hydroxy-1,4-dimethyl-5-(morpholinomethyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₃H₁₇N₃O₃ Melting Pt: 209…212 °C |
MDL Number:
MFCD05021857 CAS Number: 85843-03-4 |
Specification Test Results
Melting point | 209-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...