1-((4-Bromophenyl)sulfonyl)-4-pyridinecarboxylic acid ≥95%
Supplier: Matrix Scientific
044566-500MG
044566-5G
044566-1G
101866-498EA
806.15
USD
101866-498
101866-494
101866-496
1-((4-Bromophenyl)sulfonyl)-4-pyridinecarboxylic acid ≥95%
1-((4-Bromophenyl)sulphonyl)-4-pyridinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₄BrNO₄S Melting Pt: 224…227 °C |
MDL Number:
MFCD02331440 CAS Number: 203519-01-1 |
Specification Test Results
Melting point | 224-227°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...