3-Methoxy-3-oxo-1-propenyl-3,4,5-trimethoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
044554-1G
044554-5G
044554-500MG
101866-450EA
232.31
USD
101866-450
101866-452
101866-448
3-Methoxy-3-oxo-1-propenyl-3,4,5-trimethoxybenzenecarboxylate ≥95%
3-Methoxy-3-oxo-1-propenyl-3,4,5-trimethoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₆O₇ Melting Pt: 109…111 °C |
MDL Number:
MFCD03001266 CAS Number: 400878-22-0 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...