5-Amino-3-methyl-4-isoxazolecarboxamide ≥95%

Supplier: Matrix Scientific

044546-500MG 044546-1G 044546-5G
101866-410EA 327.69 USD
101866-410 101866-412 101866-408
5-Amino-3-methyl-4-isoxazolecarboxamide ≥95%
5-Amino-3-methyl-4-isoxazolecarboxamide

Hazard: Irritant

Formula: C₅H₇N₃O₂
Melting Pt: 196…200 °C
MDL Number: MFCD03425767
CAS Number: 35261-06-4

Order Now

Specification Test Results

Melting point 196-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR