Methyl-2,4-dioxo-4-phenylbutanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,4-dioxo-4-phenylbutanoate

044521-1G 044521-5G 044521-500MG
101866-312EA 190.77 USD
101866-312 101866-310 101866-308
Methyl-2,4-dioxo-4-phenylbutanoate ≥95%
Methyl-2,4-dioxo-4-phenylbutanoate

Hazard: Irritant

Formula: C₁₁H₁₀O₄
MW: 206.2 g/mol
Melting Pt: 59…60 °C
MDL Number: MFCD00225528
CAS Number: 20577-73-5

Order Now

Specification Test Results

Melting point 59-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR