Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(3,4-dichlorophenyl)-2,4-dioxobutanoate

044520-500MG 044520-1G 044520-5G
101866-302EA 50.77 USD
101866-302 101866-304 101866-306
Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate ≥95%
Methyl-4-(3,4-dichlorophenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₁H₈Cl₂O₄
MW: 275.09 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD03001261
CAS Number: 374679-63-7

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR