1-(4-Bromobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
044515-1G
044515-500MG
101866-282EA
467.69
USD
101866-282
101866-280
1-(4-Bromobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
1-(4-Bromobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₃BrN₂O Melting Pt: 131…133 °C |
MDL Number:
MFCD00975043 CAS Number: 338748-75-7 |
Specification Test Results
Melting point | 131-133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...