2-Hydrazino-5-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydrazinyl-5-nitropyridine

044499-1G 044499-500MG 044499-5G
101866-188EA 53.85 USD
101866-188 101866-190 101866-192
2-Hydrazino-5-nitropyridine ≥95%
2-Hydrazino-5-nitropyridine

Hazard: Irritant

Formula: C₅H₆N₄O₂
MW: 154.13 g/mol
Melting Pt: 195…196 °C
MDL Number: MFCD00118332
CAS Number: 6343-98-2

Order Now

Specification Test Results

Melting point 195-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR