Benzyl-2,4-dichloro-5-nitrophenyl ether ≥95%
Supplier: Matrix Scientific
044498-5G
044498-1G
044498-500MG
101866-186EA
1600
USD
101866-186
101866-182
101866-184
Benzyl-2,4-dichloro-5-nitrophenyl ether ≥95%
Benzyl-2,4-dichloro-5-nitrophenyl ether
Hazard: Irritant
Formula:
C₁₃H₉Cl₂NO₃ MW: 298.13 g/mol Melting Pt: 86…88 °C |
MDL Number:
MFCD08056626 CAS Number: 114109-50-1 |
Specification Test Results
Melting point | 86-88°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...