Methyl[(4-methylphenyl)methylene]ammoniumolate ≥95%

Supplier: Matrix Scientific

Synonyms: (Z)-Methyl[(4-methylphenyl)methylidene]oxidoazanium

044487-500MG 044487-5G 044487-1G
101866-124EA 278.46 USD
101866-124 101866-126 101866-128
Methyl[(4-methylphenyl)methylene]ammoniumolate ≥95%
Methyl[(4-methylphenyl)methylene]ammoniumolate

Hazard: Irritant

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00231569
CAS Number: 16089-63-7

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR