Methyl-3-hydroxyisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-hydroxyisonicotinate

044486-5G 044486-1G
101866-120EA 66.15 USD
101866-120 101866-122
Methyl-3-hydroxyisonicotinate ≥95%
Methyl 3-hydroxyisonicotinate

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.1400 g/mol
Melting Pt: 74…76 °C
MDL Number: MFCD00661299
CAS Number: 10128-72-0

Order Now

Specification Test Results

Melting point 74-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR