2-[2-(4-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid ≥95%

Supplier: Matrix Scientific

044482-500MG 044482-1G 044482-5G
101866-104EA 747.69 USD
101866-104 101866-106 101866-108
2-[2-(4-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid ≥95%
2-[2-(4-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₂S
Melting Pt: 163…165 °C
MDL Number: MFCD00663991
CAS Number: 23821-72-9

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR