5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044477-500MG 044477-1G 044477-5G
101866-076EA 927.69 USD
101866-076 101866-078 101866-074
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₃BrN₂OS
Melting Pt: 150…153 °C
MDL Number: MFCD00244934
CAS Number: 318247-52-8

Order Now

Specification Test Results

Melting point 150-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR