4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-5-methoxy-2-(methylthio)pyrimidine
044467-500MG
101866-018EA
921.54
USD
101866-018
4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine ≥97%
4-Chloro-5-methoxy-2-(methylsulfanyl)pyrimidine
Hazard: Irritant
Formula:
C₆H₇ClN₂OS MW: 190.65 g/mol Melting Pt: 73…77 °C |
MDL Number:
MFCD00223689 CAS Number: 87026-45-7 |
Specification Test Results
Melting point | 73-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...