5-(4-Chlorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
044466-5G
044466-1G
044466-500MG
101866-014EA
584.62
USD
101866-014
101866-016
101866-012
5-(4-Chlorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
5-(4-Chlorophenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₂H₇ClN₂O MW: 230.66 g/mol Melting Pt: 281…284 °C |
MDL Number:
MFCD00231524 CAS Number: 35982-98-0 |
Specification Test Results
Melting point | 281-284°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...