2-(Benzyloxy)-1-naphthaldehyde ≥95%
Supplier: Matrix Scientific
044454-500MG
044454-1G
044454-5G
101865-942EA
201.25
USD
101865-942
101865-940
101865-944
2-(Benzyloxy)-1-naphthaldehyde ≥95%
2-(Benzyloxy)-1-naphthaldehyde
Hazard: Irritant
Formula:
C₁₈H₁₄O₂ Melting Pt: 121…123 °C |
MDL Number:
MFCD00202669 CAS Number: 52805-48-8 |
Specification Test Results
Melting point | 121-123°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...