Tetrahydro-4-isoxazolol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Oxazolidin-4-ol hydrochloride

044451-1G 044451-5G 044451-500MG
101865-926EA 2129.23 USD
101865-926 101865-924 101865-922
Tetrahydro-4-isoxazolol hydrochloride ≥95%
Tetrahydro-4-isoxazolol hydrochloride

Hazard: Irritant

Formula: C₃H₈ClNO₂
MW: 125.56 g/mol
Melting Pt: 154…157 °C
MDL Number: MFCD01114602
CAS Number: 82409-18-5

Order Now

Specification Test Results

Melting point 154-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR