Succinic acid dihydrazide

Supplier: Matrix Scientific

Synonyms: Succinic Dihydrazide

036029-500MG
101840-548EA 290.77 USD
101840-548
Succinic acid dihydrazide
Succinic acid dihydrazide

Hazard: Irritant

Formula: C₄H₁₀N₄O₂
MW: 146.1478 g/mol
Melting Pt: 170…171 °C
MDL Number: MFCD00007613
CAS Number: 4146-43-4

Order Now

Specification Test Results

Melting point 170-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR