5-Chloro-6-fluoro-2,1,3-benzothiadiazole
Supplier: Matrix Scientific
002770-1G
002770-250MG
101784-618EA
56.92
USD
101784-618
101784-620
5-Chloro-6-fluoro-2,1,3-benzothiadiazole
5-Chloro-6-fluoro-2,1,3-benzothiadiazole
Hazard: Irritant
Formula:
C₆H₂ClFN₂S Melting Pt: 78 °C |
CAS Number:
175204-22-5 |
Specification Test Results
Melting point | 78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...