6-Chloro-5-fluorobenzimidazole, Technical Grade

Supplier: Matrix Scientific

002693-1G
101784-448EA 300 USD
101784-448
6-Chloro-5-fluorobenzimidazole, Technical Grade
6-Chloro-5-fluorobenzimidazole

Hazard: Irritant

Formula: C₇H₄ClFN₂
Melting Pt: 148…150 °C
MDL Number: MFCD00156132
CAS Number: 175135-04-3

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR