2-Fluoroisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoropyridine-4-carboxylic acid

002603-25G
101784-274EA 89.23 USD
101784-274
2-Fluoroisonicotinic acid ≥98%
2-Fluoroisonicotinic acid

Hazard: Irritant

Formula: C₆H₄FNO₂
MW: 141.10 g/mol
Melting Pt: 195…197 °C
MDL Number: MFCD02181194
CAS Number: 402-65-3

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR