6-Fluoro-2-methyl-4-chromanone

Supplier: Matrix Scientific

002152-1G
101783-346EA 41.54 USD
101783-346
6-Fluoro-2-methyl-4-chromanone
6-Fluoro-2-methyl-4-chromanone

Hazard: Irritant

Formula: C₁₀H₉FO₂
Melting Pt: 71 °C
MDL Number: MFCD00059072
CAS Number: 88754-96-5

Order Now

Specification Test Results

Melting point 71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR