trans-2-Methoxy-β-nitrostyrene ≥98%

Supplier: Matrix Scientific

001876-5G
101782-786EA 83.95 USD
101782-786
trans-2-Methoxy-β-nitrostyrene ≥98%
trans-2-Methoxy-β-nitrostyrene

Hazard: Irritant/Refrigerate

Formula: C₉H₉NO₃
Melting Pt: 46…48 °C
MDL Number: MFCD00070647
CAS Number: 3316-24-3

Order Now

Specification Test Results

Melting point 46-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR