4-Amino-3-ethylbenzonitrile ≥97%
Supplier: Matrix Scientific
001673-5G
001673-1G
101782-238EA
61.54
USD
101782-238
101782-240
4-Amino-3-ethylbenzonitrile ≥97%
4-Amino-3-ethylbenzonitrile
Hazard: Irritant/Irritant-Harmful
Formula:
C₉H₁₀N₂ Melting Pt: 58…61 °C |
MDL Number:
MFCD00041431 CAS Number: 170230-87-2 |
Specification Test Results
Melting point | 58-61°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...