3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol ≥97%

Supplier: Matrix Scientific

001574-500MG
101781-992EA 638.46 USD
101781-992
3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol ≥97%
3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol

Hazard: Irritant

Formula: C₁₇H₈F₁₂O
Melting Pt: 148 °C
MDL Number: MFCD00042475
CAS Number: 87901-76-6

Order Now

Specification Test Results

Melting point 148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR