Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

2-Hydroxy-5-methylisophthalaldehyde

Supplier: Matrix Scientific


001506-1G 001506-5G
101781-842EA 77.15 USD InStock
101781-842 101781-844
2-Hydroxy-5-methylisophthalaldehyde
2-Hydroxy-5-methylisophthalaldehyde
CAS: 7310-95-4
MDL Number: MFCD00016596
Molecular Formula: C9H8O3
Molecular Weight: 164,16
Hazard: Irritant


Formula: C₉H₈O₃
Melting Pt: 128…130 °C
MDL Number: MFCD00016596
CAS Number: 7310-95-4
EINECS: 230-768-4
Order Now
Specification Test Results
Melting point 130°C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty