4'-Nitro-2'-(trifluoromethyl)acetanilide ≥97%
Supplier: Matrix Scientific
001350-1G
001350-5G
101781-456EA
87.69
USD
101781-456
101781-454
4'-Nitro-2'-(trifluoromethyl)acetanilide ≥97%
4'-Nitro-2'-(trifluoromethyl)acetanilide
Hazard: Irritant
Formula:
C₉H₇F₃N₂O₃ Melting Pt: 144…146 °C |
MDL Number:
MFCD00052846 CAS Number: 395-68-6 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...