7-Amino-4-(trifluoromethyl)coumarin ≥99%

Supplier: Matrix Scientific

Synonyms: 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one

001336-1G
101781-422EA 46.15 USD
101781-422
7-Amino-4-(trifluoromethyl)coumarin ≥99%
7-Amino-4-(trifluoromethyl)coumarin

TCA listed: T
Hazard: Irritant

Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
Melting Pt: 220…224 °C
MDL Number: MFCD00006858
CAS Number: 53518-15-3

Order Now

Specification Test Results

Melting point 220-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR