4-Bromo-2,5-difluorobenzenesulfonamide

Supplier: Matrix Scientific

001283-1G 001283-5G
101781-308EA 28.75 USD
101781-308 101781-310
4-Bromo-2,5-difluorobenzenesulfonamide
4-Bromo-2,5-difluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₄BrF₂NO₂S
Melting Pt: 167…169 °C
MDL Number: MFCD01569463
CAS Number: 214209-98-0

Order Now

Specification Test Results

Melting point 167-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR