2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile ≥97%

Supplier: Matrix Scientific

001142-1G
101781-040EA 28.75 USD
101781-040
2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile ≥97%
2,6-Bis(2,2,2-trifluoroethoxy)benzonitrile

Hazard: Irritant

Formula: C₁₁H₁₁F₆NO₂
Melting Pt: 115…117 °C
MDL Number: MFCD00109080
CAS Number: 93624-57-8

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR