2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Difluoromethoxy)phenacyl bromide

001064-1G
101780-896EA 70.77 USD
101780-896
2-Bromo-4'-(difluoromethoxy)acetophenone ≥98%
2-Bromo-4'-(difluoromethoxy)acetophenone

Hazard: Irritant

Formula: C₉H₇BrF₂O₂
Melting Pt: 66…68 °C
MDL Number: MFCD00052200
CAS Number: 141134-24-9

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR