    Sort by:
  • Search Within Results
Spouting Cylinders
https://us.vwr.com/stibo/smallweb/std.lang.all/69/47/20236947.jpg
VWR offers spouting cylinders for the demonstration of Torricelli's law. Clear or colored acrylic cylinders feature three vertically aligned spouting nozzles or holes with an optional filler hose barb at the top to maintain a consistent water depth. Fill the cylinder to visually demonstrate that water pressure changes significantly with depth. Cylinders are appropriate for small group demonstration and experimentation.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (470303-934)
Supplier:  United Scientific Supplies
Description:   Demonstrate Torricelli’s theorem to show the relationship between water pressure and depth.